missing translation for 'onlineSavingsMsg'
Learn More

A 784168, Tocris Bioscience™

Potent and selective TRPV1 antagonist

Brand:  RD Systems (Bio-Techne) 4319/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13444554

  • £183.48 / 10mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent TRPV1 antagonist (IC50 = 25 nM for inhibition of TRPV1 activation by 50 nM capsaicin). Displays no activity against a range of receptors, including TRPA1, GABA, opioid, and purinergic receptors.
Specifications

Chemical Identifiers

824982-41-4
479.397
unii-lb160j37hd, 3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, n-4-trifluoromethanesulfonylphenyl-3'-trifluoromethyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, 1 2h ,2'-bipyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl, 1-3-trifluoromethyl pyridin-2-yl-n-4-trifluoromethylsulfonyl phenyl-1,2,3,6-tetrahydropyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-1 2h ,2'-bipyridine-4-carboxamide, n-4-trifluoromethane sulfonylphenyl-1-3-trifluoromethyl pyridin-2-yl-1,2,3,6-tetrahydropyridine-4-carboxamide
1-[3-(trifluoromethyl)pyridin-2-yl]-N-[4-(trifluoromethylsulfonyl)phenyl]-3,6-dihydro-2H-pyridine-4-carboxamide
C19H15F6N3O3S
SDUAWRFBHRAFBM-UHFFFAOYSA-N
11420211
C1CN(CC=C1C(=O)NC2=CC=C(C=C2)S(=O)(=O)C(F)(F)F)C3=C(C=CC=N3)C(F)(F)F

Specifications

A 784168
824982-41-4
unii-lb160j37hd, 3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, n-4-trifluoromethanesulfonylphenyl-3'-trifluoromethyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, 1 2h ,2'-bipyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl, 1-3-trifluoromethyl pyridin-2-yl-n-4-trifluoromethylsulfonyl phenyl-1,2,3,6-tetrahydropyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-1 2h ,2'-bipyridine-4-carboxamide, n-4-trifluoromethane sulfonylphenyl-1-3-trifluoromethyl pyridin-2-yl-1,2,3,6-tetrahydropyridine-4-carboxamide
C1CN(CC=C1C(=O)NC2=CC=C(C=C2)S(=O)(=O)C(F)(F)F)C3=C(C=CC=N3)C(F)(F)F
479.397
479.4
10mg
C19H15F6N3O3S
SDUAWRFBHRAFBM-UHFFFAOYSA-N
1-[3-(trifluoromethyl)pyridin-2-yl]-N-[4-(trifluoromethylsulfonyl)phenyl]-3,6-dihydro-2H-pyridine-4-carboxamide
11420211
>99%
Videos