missing translation for 'onlineSavingsMsg'
Learn More

A 77636 hydrochloride, Tocris Bioscience™

Potent and selective D1-like agonist; orally active

Brand:  Tocris (Bio-Techne) 170110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448663

  • £198.45 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective dopamine D1-like receptor agonist (pEC50 values are 8.97 and < 5 for D1-like and D2-like receptors respectively). Displays anti-Parkinsonian activity following oral administration in vivo. Exhibits 11-fold cell type bias over dopamine in a functional assay in U2 cells.
Specifications

Chemical Identifiers

145307-34-2
365.898
(1R-cis)-1-(Aminomethyl)-3,4-dihydro-3-tricyclo[3.3.1.13,7]dec-1-yl-[1H]-2-benzopyran-5,6-diol hydrochloride
3-(1-adamantyl)-1-(aminomethyl)-3,4-dihydro-1H-isochromene-5,6-diol;hydrochloride
C20H28ClNO3
BWHPNJVKFAPVOG-UHFFFAOYSA-N
16069211
C1C2CC3CC1CC(C2)(C3)C4CC5=C(C=CC(=C5O)O)C(O4)CN.Cl

Specifications

10mg
145307-34-2
(1R-cis)-1-(Aminomethyl)-3,4-dihydro-3-tricyclo[3.3.1.13,7]dec-1-yl-[1H]-2-benzopyran-5,6-diol hydrochloride
C1C2CC3CC1CC(C2)(C3)C4CC5=C(C=CC(=C5O)O)C(O4)CN.Cl
365.898
365.9
A 77636 hydrochloride
C20H28ClNO3
BWHPNJVKFAPVOG-UHFFFAOYSA-N
3-(1-adamantyl)-1-(aminomethyl)-3,4-dihydro-1H-isochromene-5,6-diol;hydrochloride
16069211
>98%
Videos
Documents