missing translation for 'onlineSavingsMsg'
Learn More

A 68930 hydrochloride, Tocris Bioscience™

Potent, selective D1-like agonist

Brand:  Tocris (Bio-Techne) 153450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449623

  • £601.23 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective D1-like dopamine receptor agonist (EC50 values are 2.1 and 3910 nM for D1-like and D2-like receptors respectively). Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

130465-39-3
307.774
a 68930 hydrochloride, a-68930 hydrochloride, cis-+/--1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride, cis-1-aminomethyl-3-phenylisochroman-5,6-diol hydrochloride, 1r,3s-1-aminomethyl-3-phenyl-3,4-dihydro-1h-2-benzopyran-5,6-diol hydrochloride, a-68930 hydrochloride hplc , powder, cis-1-aminomethyl-3-phenyl-3,4-dihydro-5,6-dihydroxy-1h-2-benzopyran hydrochloride, cis-inverted exclamation marka-1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride
(1R,3S)-1-(aminomethyl)-3-phenyl-3,4-dihydro-1H-isochromene-5,6-diol;hydrochloride
C16H18ClNO3
PQPGUUQPTSMLKU-YYLIZZNMSA-N
9904672
C1C(OC(C2=C1C(=C(C=C2)O)O)CN)C3=CC=CC=C3.Cl

Specifications

50mg
130465-39-3
a 68930 hydrochloride, a-68930 hydrochloride, cis-+/--1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride, cis-1-aminomethyl-3-phenylisochroman-5,6-diol hydrochloride, 1r,3s-1-aminomethyl-3-phenyl-3,4-dihydro-1h-2-benzopyran-5,6-diol hydrochloride, a-68930 hydrochloride hplc , powder, cis-1-aminomethyl-3-phenyl-3,4-dihydro-5,6-dihydroxy-1h-2-benzopyran hydrochloride, cis-inverted exclamation marka-1-aminomethyl-3,4-dihydro-3-phenyl-1h-2-benzopyran-5,6-diol hydrochloride
C1C(OC(C2=C1C(=C(C=C2)O)O)CN)C3=CC=CC=C3.Cl
307.774
307.78
A 68930 hydrochloride
C16H18ClNO3
PQPGUUQPTSMLKU-YYLIZZNMSA-N
(1R,3S)-1-(aminomethyl)-3-phenyl-3,4-dihydro-1H-isochromene-5,6-diol;hydrochloride
9904672
>98%
Videos
Documents