missing translation for 'onlineSavingsMsg'
Learn More

A 61603 hydrobromide, Tocris Bioscience™

α1A agonist

Brand:  Tocris (Bio-Techne) 105210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438393

  • £180.81 / 10mg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent α-adrenoceptor agonist that is at least 35-fold more potent at α1A than at α1B or α1D sites. Induces dose response increases in spontaneous Ca2+ transients in rat ventricular myocytes in vitro (EC50 = 6.9 nmol/L). Also available as part of the α1-Adrenoceptor Tocriset™ .
Specifications

Chemical Identifiers

107756-30-9
390.296
a 61603 hydrobromide, n-5-4,5-dihydro-1h-imidazol-2-yl-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl methanesulfonamide hydrobromide
N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide;hydrobromide
C14H20BrN3O3S
LRFLWCZMTGTUEP-UHFFFAOYSA-N
9865178
CS(=O)(=O)NC1=C(C=CC2=C1CCCC2C3=NCCN3)O.Br

Specifications

10mg
107756-30-9
a 61603 hydrobromide, n-5-4,5-dihydro-1h-imidazol-2-yl-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl methanesulfonamide hydrobromide
CS(=O)(=O)NC1=C(C=CC2=C1CCCC2C3=NCCN3)O.Br
390.296
390.29
A 61603 hydrobromide
C14H20BrN3O3S
LRFLWCZMTGTUEP-UHFFFAOYSA-N
N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide;hydrobromide
9865178
>98%
Videos
Documents