missing translation for 'onlineSavingsMsg'
Learn More

A 484954, Tocris Bioscience™

CaM kinase III (eEF-2 kinase) inhibitor

Brand:  RD Systems (Bio-Techne) 4483/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444824

  • £764.40 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of eukaryotic elongation factor-2 (eEF-2) kinase (IC50 = 0.28 μM in enzymatic assay). Reduces eEF-2 phosphorylation with little effect on cancer cell growth.
Specifications

Chemical Identifiers

142557-61-7
289.295
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxo-1,2,3,4-tetrahydropyrido 2,3-d pyrimidine-6-carboxamide, 7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido 2,3-d pyrimidine-6-carboxamide, 7-amino-1-cyclopropyl-3-ethyl-1,2,3,4-tetrahydro-2,4-dioxopyrido 2,3-d pyrimidine-6-carboxamide, kinome_3822
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
C13H15N5O3
HWODCHXORCTEGU-UHFFFAOYSA-N
14998470
CCN1C(=O)C2=CC(=C(N=C2N(C1=O)C3CC3)N)C(=O)N

Specifications

50mg
142557-61-7
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxo-1,2,3,4-tetrahydropyrido 2,3-d pyrimidine-6-carboxamide, 7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido 2,3-d pyrimidine-6-carboxamide, 7-amino-1-cyclopropyl-3-ethyl-1,2,3,4-tetrahydro-2,4-dioxopyrido 2,3-d pyrimidine-6-carboxamide, kinome_3822
CCN1C(=O)C2=CC(=C(N=C2N(C1=O)C3CC3)N)C(=O)N
289.295
289.29
A 484954
C13H15N5O3
HWODCHXORCTEGU-UHFFFAOYSA-N
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
14998470
>98%
Videos
Documents