missing translation for 'onlineSavingsMsg'
Learn More

A 438079 hydrochloride, Tocris Bioscience™

Competitive P2X7 antagonist

Brand:  Tocris (Bio-Techne) 297250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462164

  • £600.29 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Competitive P2X7 receptor antagonist (pIC50 = 6.9 for the inhibition of Ca2+ influx in the human recombinant P2X7 cell line). Devoid of activity at other P2 receptors (IC50 >> 10 μM). Possesses antinociceptive activity in models of neuropathic pain in vivo. Inhibits methamphetamine-induced microglial migration and phagocytosis in vitro.
Specifications

Chemical Identifiers

899431-18-6
342.608
a 438079 hydrochloride, a 438079 hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1,2,3,4-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl, c13h9cl2n5.clh, hcl, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl tetrazol-1-yl methyl pyridine;hydrochloride
3-[[5-(2,3-dichlorophenyl)tetrazol-1-yl]methyl]pyridine;hydrochloride
C13H10Cl3N5
MBTJFFMIPPMRGR-UHFFFAOYSA-N
11688742
C1=CC(=C(C(=C1)Cl)Cl)C2=NN=NN2CC3=CN=CC=C3.Cl

Specifications

A 438079 hydrochloride
899431-18-6
a 438079 hydrochloride, a 438079 hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1,2,3,4-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl, c13h9cl2n5.clh, hcl, 3-5-2,3-dichlorophenyl-1h-tetrazol-1-yl methyl pyridine hydrochloride, 3-5-2,3-dichlorophenyl tetrazol-1-yl methyl pyridine;hydrochloride
C1=CC(=C(C(=C1)Cl)Cl)C2=NN=NN2CC3=CN=CC=C3.Cl
342.608
342.61
50mg
C13H10Cl3N5
MBTJFFMIPPMRGR-UHFFFAOYSA-N
3-[[5-(2,3-dichlorophenyl)tetrazol-1-yl]methyl]pyridine;hydrochloride
11688742
>98%
Videos