missing translation for 'onlineSavingsMsg'
Learn More

A 412997 dihydrochloride, Tocris Bioscience™

Selective D4 agonist

Brand:  RD Systems (Bio-Techne) 4552/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13434924

  • £145.53 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective agonist for the dopamine D4 receptor (Ki values are 7.9 and 12.1 nM for human D4 and rat D4 receptors). Displays no affinity (<1000 nM) for other dopamine receptors. Shows efficacy in rat models of ADHD and short-term memory.
Specifications

Chemical Identifiers

1347744-96-0
382.329
a-412997 dihydrochloride, a 412997 dihydrochloride, 2-4-pyridin-2-yl piperidin-1-yl-n-m-tolyl acetamide dihydrochloride, n-3-methylphenyl-4-2-pyridinyl-1-piperidineacetamide dihydrochloride, n-3-methylphenyl-2-4-pyridin-2-yl piperidin-1-yl acetamide dihydrochloride
N-(3-methylphenyl)-2-(4-pyridin-2-ylpiperidin-1-yl)acetamide;dihydrochloride
C19H25Cl2N3O
KIGNSSHMCNBHQX-UHFFFAOYSA-N
90488953
CC1=CC(=CC=C1)NC(=O)CN2CCC(CC2)C3=CC=CC=N3.Cl.Cl

Specifications

10mg
1347744-96-0
a-412997 dihydrochloride, a 412997 dihydrochloride, 2-4-pyridin-2-yl piperidin-1-yl-n-m-tolyl acetamide dihydrochloride, n-3-methylphenyl-4-2-pyridinyl-1-piperidineacetamide dihydrochloride, n-3-methylphenyl-2-4-pyridin-2-yl piperidin-1-yl acetamide dihydrochloride
CC1=CC(=CC=C1)NC(=O)CN2CCC(CC2)C3=CC=CC=N3.Cl.Cl
382.329
382.33
A 412997 dihydrochloride
C19H25Cl2N3O
KIGNSSHMCNBHQX-UHFFFAOYSA-N
N-(3-methylphenyl)-2-(4-pyridin-2-ylpiperidin-1-yl)acetamide;dihydrochloride
90488953
>98%
Videos
Documents