missing translation for 'onlineSavingsMsg'
Learn More

A 350619 hydrochloride, Tocris Bioscience™

Soluble guanylyl cyclase (sGC) activator

Brand:  Tocris (Bio-Techne) 275350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460524

  • £426.42 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Activator of soluble guanylyl cyclase (sGC). Activates basal sGC and synergistically activates sGC in the presence of NO. Relaxes cavernosum smooth muscle in vitro and induces penile erections in rats. Induces a sustained increase in skin blood flow in vivo.
Specifications

Chemical Identifiers

1217201-17-6
425.412
a 350619 hydrochloride, 3-2-4-chlorophenyl thiophenyl-n-4-dimethylamino butyl-2-propenamide hydrochloride, 3-2-4-chlorophenyl thiophenyl-n-4-dimethylamino butyl-2-propenamidehcl
(E)-3-[2-(4-chlorophenyl)sulfanylphenyl]-N-[4-(dimethylamino)butyl]prop-2-enamide;hydrochloride
C21H26Cl2N2OS
PDVBHWZPRQFKJS-KYIGKLDSSA-N
56924282
CN(C)CCCCNC(=O)C=CC1=CC=CC=C1SC2=CC=C(C=C2)Cl.Cl

Specifications

A 350619 hydrochloride
1217201-17-6
a 350619 hydrochloride, 3-2-4-chlorophenyl thiophenyl-n-4-dimethylamino butyl-2-propenamide hydrochloride, 3-2-4-chlorophenyl thiophenyl-n-4-dimethylamino butyl-2-propenamidehcl
CN(C)CCCCNC(=O)C=CC1=CC=CC=C1SC2=CC=C(C=C2)Cl.Cl
425.412
425.41
50mg
C21H26Cl2N2OS
PDVBHWZPRQFKJS-KYIGKLDSSA-N
(E)-3-[2-(4-chlorophenyl)sulfanylphenyl]-N-[4-(dimethylamino)butyl]prop-2-enamide;hydrochloride
56924282
>99%
Videos