missing translation for 'onlineSavingsMsg'
Learn More

A 205804, Tocris Bioscience™

Selective inhibitor of E-selectin and ICAM-1 expression

Brand:  Tocris (Bio-Techne) 252450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431394

  • £646.80 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of E-selectin and ICAM-1 expression (IC50 values are 20, 25 and > 1000 nM for TNF-α-induced E-selectin, ICAM-1 and VCAM-1 expression respectively). Effective inhibitor of cell-cell adhesion in an in vitro flow experiment.
Specifications

Chemical Identifiers

251992-66-2
300.394
4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide
4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide
C15H12N2OS2
QQGWEXFLMJGCAL-UHFFFAOYSA-N
9839311
CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N

Specifications

50mg
251992-66-2
4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide, unii-tp7tb1sszd, tp7tb1sszd, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide, 4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide, thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio, 4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca, 4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide
CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N
300.394
300.39
A 205804
C15H12N2OS2
QQGWEXFLMJGCAL-UHFFFAOYSA-N
4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide
9839311
>98%
Videos
Documents