missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopurine, 98%, ACROS Organics™

Brand:  Acros Organics 276562500

Code : 29

Additional Details : CAS Number : 452-06-2 Weight : 0.05025kg


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10217220

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Aminopurine
97.5
≥97.5%
C5H5N5
1H-Purine-2-amine
452-06-2
100.0
98%
135.13

Specifications

2-Aminopurine
97.5
Authentic
Amorphous Powder
98%
280° to 284°C
Light beige
250mg
452-06-2
100.0
Glass bottle
≥97.5%
C5H5N5
135.13
26, 414
1H-Purine-2-amine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Aminopurine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS