Learn More
Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates
Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.
Brand:  Eppendorf™ 0030132556
Code : 96
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 10033373
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Improved well-to-well tolerance
  • Semi-skirt for labelling
  • Extremely thin-walled for optimum heat transfer into the sample
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing and reduced cross-contamination risk
  • 250μL well volume (when used with cap strips)
Specifications
Specifications
96-Well PCR Plate
Yes
121°C
121°C
Conical
Yes
For PCR techniques
Polycarbonate
Quantitative protein analysis
96 Wells, Semi-Skirted
Videos