missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ 96-Well Sterile Polypropylene U-Bottom Microplates
Pipette, control and recover samples with this polypropylene microplate. Eppendorf™ 96-Well Sterile Polypropylene U-Bottom Microplates are ideal for assays that require high resistance against temperature or solvents.
Brand: Eppendorf™ 0030606205
Code : 96
Additional Details : Weight : 8.05500kg
Description
- Unmatched, transparent polypropylene (PP) for improved sample visibility
- OptiTrack™ Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax™ well design: Optimized well geometry for minimal remaining volume and excellent mixing properties
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- g-Safe™: exceptional centrifugation stability up to 6,000 × g
- Polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers or biocides – Materials which could have a negative effect on bioassays
- Guaranteed functionality from -86 to +100°C
- Autoclavable (121°C, 20 min.)
Specifications
Standard | |
White, Clear Bottom | |
96 | |
Round Bottom (U-well) | |
White |
8 x 12 | |
Polypropylene | |
Sterile | |
240/Pk. | |
Clear |