missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Non Binding Microplates
Features long-term surface stability without degradation or leaching.
POA
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Autoclavable | Not Autoclavable |
Color | Clear |
Material | Polystyrene |
Description
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Significantly increase assay sensitivity
Specifications
Standard | |
Not Autoclavable | |
Polystyrene | |
Non-sterile | |
40/Cs. |
8 x 12 | |
Clear | |
96 | |
40 | |
Chimney |