missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex 95799 Insole For Uvex 1 Sport - Width 12
Comfortable climatic insole with antistatic properties for Uvex 1 Sport
£17.10
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Uvex 1 Sport |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17208991
|
Uvex
9579935 |
35 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17218991
|
Uvex
9579936 |
36 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17228991
|
Uvex
9579937 |
37 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17238991
|
Uvex
9579938 |
38 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17248991
|
Uvex
9579939 |
39 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17258991
|
Uvex
9579940 |
40 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17268991
|
Uvex
9579941 |
41 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17278991
|
Uvex
9579942 |
42 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17288991
|
Uvex
9579943 |
43 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17298991
|
Uvex
9579944 |
44 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17209001
|
Uvex
9579945 |
45 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17219001
|
Uvex
9579946 |
46 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17229001
|
Uvex
9579947 |
47 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17239001
|
Uvex
9579948 |
48 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17249001
|
Uvex
9579949 |
49 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17259001
|
Uvex
9579951 |
51 |
£17.10
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9579.9
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 Sport | |
No | |
Wide width | |
No |