missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex 95289 High Tune-Up Insole For Uvex 1 G2 - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
£19.80
Specifications
Color | Black, Orange |
---|---|
For Use With (Equipment) | Uvex 1 G2 |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17258141
|
Uvex
9528935 |
35 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17268141
|
Uvex
9528936 |
36 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17278141
|
Uvex
9528937 |
37 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17288141
|
Uvex
9528938 |
38 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17298141
|
Uvex
9528939 |
39 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17208151
|
Uvex
9528940 |
40 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17218151
|
Uvex
9528941 |
41 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17228151
|
Uvex
9528942 |
42 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17238151
|
Uvex
9528943 |
43 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17248151
|
Uvex
9528944 |
44 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17258151
|
Uvex
9528945 |
45 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17268151
|
Uvex
9528946 |
46 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17278151
|
Uvex
9528947 |
47 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17288151
|
Uvex
9528948 |
48 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17298151
|
Uvex
9528949 |
49 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17208161
|
Uvex
9528950 |
50 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17218161
|
Uvex
9528951 |
51 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17228161
|
Uvex
9528952 |
52 |
£19.80
1 pair |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Uvex Tune-Up Comfortable High Insole 95289
- Antistatic insole concept for use in safety footwear
- Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
- Good cushioning
- Relief of pressure marks
- Non-irritating, comfortable surface structure
- Breathable and absorbs moisture
- Antibacterial
- Antistatic
- Useable for ESD-footwear
- Uvex 1 G2
- Article No. 95289
- Standard EN ISO 20345:2011
- Sizes: 35 to 52
Specifications
Black, Orange | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 G2 | |
No | |
Standard width | |
No |