missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex 95283 High Tune-Up Insole For Uvex 1 and 2 - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
£19.80
Specifications
Color | Black, Orange |
---|---|
For Use With (Equipment) | Uvex 1 and 2 |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17278031
|
Uvex
9528335 |
35 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17288031
|
Uvex
9528336 |
36 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17298031
|
Uvex
9528337 |
37 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17208041
|
Uvex
9528338 |
38 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17218041
|
Uvex
9528339 |
39 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17228041
|
Uvex
9528340 |
40 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17238041
|
Uvex
9528341 |
41 |
£19.80
1 pair |
In Stock
Log in to see stock. |
|||||
17248041
|
Uvex
9528342 |
42 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17258041
|
Uvex
9528343 |
43 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17268041
|
Uvex
9528344 |
44 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17278041
|
Uvex
9528345 |
45 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17288041
|
Uvex
9528346 |
46 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17298041
|
Uvex
9528347 |
47 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17208051
|
Uvex
9528348 |
48 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17218051
|
Uvex
9528349 |
49 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17228051
|
Uvex
9528350 |
50 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17238051
|
Uvex
9528351 |
51 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
17248051
|
Uvex
9528352 |
52 |
£19.80
1 pair |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
Uvex Tune-Up Comfortable High Insole 95283
- Antistatic insole concept for use in safety footwear
- Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
- Good cushioning
- Relief of pressure marks
- Non-irritating, comfortable surface structure
- Breathable and absorbs moisture
- Antibacterial
- Antistatic
- Useable for ESD-footwear
- Uvex 1, Uvex 2, Uvex 1 Duo, Uvex Xenova™ Light
- Article No. 95283
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 and 2 | |
No | |
Standard width | |
No |