missing translation for 'onlineSavingsMsg'
Learn More

Uvex 94909 Comfortable Climate Insole For Uvex G2 - Width 12

Comfortable climatic insole with antistatic properties for Uvex 1 G2

£17.10

Specifications

Color Black
For Use With (Equipment) Uvex G2
Material Microvelour
Disposable No
Gender Unisex
View More Specs

Products 16
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17277441
View Documents Promotion Details
Uvex
9490935
35
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17287441
View Documents Promotion Details
Uvex
9490936
36
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17297441
View Documents Promotion Details
Uvex
9490937
37
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17207451
View Documents Promotion Details
Uvex
9490938
38
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17217451
View Documents Promotion Details
Uvex
9490939
39
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17227451
View Documents Promotion Details
Uvex
9490940
40
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17237451
View Documents Promotion Details
Uvex
9490941
41
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17247451
View Documents Promotion Details
Uvex
9490942
42
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17257451
View Documents Promotion Details
Uvex
9490943
43
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17267451
View Documents Promotion Details
Uvex
9490944
44
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17277451
View Documents Promotion Details
Uvex
9490945
45
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17287451
View Documents Promotion Details
Uvex
9490946
46
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17297451
View Documents Promotion Details
Uvex
9490947
47
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17207461
View Documents Promotion Details
Uvex
9490948
48
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17217461
View Documents Promotion Details
Uvex
9490949
49
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
17227461
View Documents Promotion Details
Uvex
9490951
51
£17.10
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
Description

Description

  • Full-surface foot shock absorption
  • Highly breathable, moisture-absorbent and quickly re-drying material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9490.9
  • Sizes: 35 to 52
Specifications

Specifications

Black
Microvelour
Unisex
Yes
Uvex G2
No
Wide width
No
Videos
SDS
Documents

Documents

Product Certifications