Learn More
Uvex™ 9174 Skylite Safety Glasses
8.65 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Lens Tint:
Clear
Gray
Frame Color:
Blue
Red
Description
- Safety glasses are extremely lightweight, combining comfort with effective eye protection
- Certified in line with EN 166 (personal eye protection)
- Ultradura lens coating is extremely scratch-resistant on the interior and exterior
- Integrated brow and cheek guards for increased wearer protection
- Spectacles protect against liquid droplets/splashes, and large dust particles
- Flexible, closed-pore soft components of frame adapt to individual face contours
- Easy cleaning process due to separate lens and frame concept
- 5-stage side arm length adjustment ensures optimum fit
Specifications
Lens Type | Ultradura |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Frame Color | Red |
Model | 9174 Skylite |
Lens Material | Polycarbonate |