missing translation for 'onlineSavingsMsg'
Learn More

9-(Trifluoroacetyl)anthracene, 98%, Thermo Scientific™

Catalog Number p-204617
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
53531-31-0
MFCD00001258
MNCMBBIFTVWHIP-UHFFFAOYSA-N
9-trifluoroacetylanthracene, 9-anthryl trifluoromethyl ketone, 1-anthracen-9-yl-2,2,2-trifluoroethanone, 9-trifluoroacetyl anthracene, 1-9-anthryl-2,2,2-trifluoroethan-1-one, 1-9-anthryl-2,2,2-trifluoroethanone, 9-anthryl, pubchem16290, acmc-209l8i, bidd:gt0598
104525
1-anthracen-9-yl-2,2,2-trifluoroethanone
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F
This item is not returnable. View return policy
53531-31-0
MFCD00001258
MNCMBBIFTVWHIP-UHFFFAOYSA-N
9-trifluoroacetylanthracene, 9-anthryl trifluoromethyl ketone, 1-anthracen-9-yl-2,2,2-trifluoroethanone, 9-trifluoroacetyl anthracene, 1-9-anthryl-2,2,2-trifluoroethan-1-one, 1-9-anthryl-2,2,2-trifluoroethanone, 9-anthryl, pubchem16290, acmc-209l8i, bidd:gt0598
104525
1-anthracen-9-yl-2,2,2-trifluoroethanone
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 83°C to 86°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H9F3O
Quantity 1 g
Molecular Weight (g/mol) 274.24
Formula Weight 274.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 9-(Trifluoroacetyl)anthracene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'