missing translation for 'onlineSavingsMsg'
Learn More

9-(Trifluoroacetyl)anthracene, 98%, ACROS Organics™

Brand:  Acros Organics 203470050

Code : 29

Additional Details : CAS Number : 53531-31-0 Weight : 0.06000kg

Packaging Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10160262

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

9-(Trifluoroacetyl)anthracene
53531-31-0
100.0
98%
274.24
9-Anthryl trifluoromethyl ketone
98%
97.5
≥97.5%
C16H9F3O
MFCD00001258

Specifications

9-(Trifluoroacetyl)anthracene
83° to 86°C
98%
97.5
Authentic
Powder
98%
274.24
9-Anthryl trifluoromethyl ketone
Yellow
5g
53531-31-0
100.0
Glass Bottle
≥97.5%
C16H9F3O
MFCD00001258
Videos
SDS