missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenylcarbazole, 99%, Thermo Scientific Chemicals
74.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
9-Phenylcarbazole has been used in the electrochemical synthesis of poly(9-phenylcarbazole) films via direct anodic oxidation in mixed electrolytes of boron trifluoride diethyl etherate and sulfuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Phenylcarbazole has been used in the electrochemical synthesis of poly(9-phenylcarbazole) films via direct anodic oxidation in mixed electrolytes of boron trifluoride diethyl etherate and sulfuric acid.
Solubility
Slightly soluble in water.
Notes
Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
9-Phenylcarbazole has been used in the electrochemical synthesis of poly(9-phenylcarbazole) films via direct anodic oxidation in mixed electrolytes of boron trifluoride diethyl etherate and sulfuric acid.
Solubility
Slightly soluble in water.
Notes
Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 95°C to 97°C |
Quantity | 5 g |
Synonym | n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 9-phenylcarbazole |
Formula Weight | 243.3 |
Percent Purity | 99% |
Chemical Name or Material | 9-Phenylcarbazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'