missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Phenylcarbazole-3-boronic acid, 98%

Brand:  Alfa Aesar™ H64059.03

Code : NEW

Additional Details : CAS Number : 854952-58-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15411089

  • £41.50 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Description

Description

Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

9-Phenylcarbazole-3-boronic acid
287.13
10566519
854952-58-2
MFCD12196936

Specifications

9-Phenylcarbazole-3-boronic acid
854952-58-2
98%
MFCD12196936
1g
Solid
287.13
10566519
Videos
SDS