missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Phenylanthracene, 98%

Brand:  Alfa Aesar™ H64419.06

Code : NEW

Additional Details : CAS Number : 602-55-1 Weight : 0.00500kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15451619

  • £65.50 / 5g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

9-Phenylanthracene
254.33
UN3077
602-55-1
MFCD00001252
1910570

Specifications

9-Phenylanthracene
417°C (lit.)
153° to 155°C (lit.)
602-55-1
98%
MFCD00001252
1910570
Light Sensitive
Off-white to pale brown
5g
Solid
254.33
UN3077
Videos
SDS