missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog Number p-3736515
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28920-43-6
C15H11ClO2
258.69
MFCD00001138
IRXSLJNXXZKURP-UHFFFAOYSA-N
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
34367
9H-fluoren-9-ylmethyl carbonochloridate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl
This item is not returnable. View return policy
28920-43-6
C15H11ClO2
258.69
MFCD00001138
IRXSLJNXXZKURP-UHFFFAOYSA-N
9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
34367
9H-fluoren-9-ylmethyl carbonochloridate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 63.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4190
Solubility Information Solubility in water: decomposes in water.,acetone
Formula Weight 258.69
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethyl chloroformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'