Learn More
9-Fluorenylmethyl chloroformate, 98+%, Thermo Scientific Chemicals
Description
Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Base sensitive amino protecting group for solid-phase peptide synthesis9-Fluorenylmethyl chloroformate is used in capillary electrophoresis. It acts as a reagent in the precolumn derivatization of amines for HPLC and fluorescent detection. Further, it is used for derivatizing amino acids for HPLC analysis. In addition to this, it is used to prepare N-Fmoc amino acids for solid-phase peptide synthesis and oligonucleotide synthesis.
Solubility
Soluble in dioxane.
Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols and amines.
Specifications
Melting Point | 60°C to 64°C |
Assay Percent Range | 98% |
Quantity | 1 g |
UN Number | UN2923 |
Beilstein | 2279177 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dioxane. |
Formula Weight | 258.7 |
Percent Purity | 98% |
Chemical Name or Material | 9-Fluorenylmethyl chloroformate |
RUO – Research Use Only