missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020827
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
486-25-9
C13H8O
180.206
MFCD00001141
YLQWCDOCJODRMT-UHFFFAOYSA-N
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
10241
CHEBI:17922
fluoren-9-one
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
This item is not returnable. View return policy
486-25-9
C13H8O
180.206
MFCD00001141
YLQWCDOCJODRMT-UHFFFAOYSA-N
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
10241
CHEBI:17922
fluoren-9-one
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

Solubility
Soluble in water, alcohol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 82°C to 85°C
Density 0.9
Boiling Point 342°C
Flash Point 163°C (325°F)
Assay Percent Range 98%
Quantity 100 g
Beilstein 1636531
Solubility Information Soluble in water,alcohol,acetone,ether and benzene.
Formula Weight 180.21
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'