missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenemethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3757932
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24324-17-2
C14H12O
196.25
MFCD00001139
XXSCONYSQQLHTH-UHFFFAOYSA-N
9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
90466
OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
24324-17-2
C14H12O
196.25
MFCD00001139
XXSCONYSQQLHTH-UHFFFAOYSA-N
9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
90466
OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 104°C to 107°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 06, III, 3507
Fieser 05,308; 03,145; 04,237; 13,134
IUPAC Name (9H-fluoren-9-yl)methanol
Formula Weight 196.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenemethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'