missing translation for 'onlineSavingsMsg'
Learn More

9-Ethynyl-9-fluorenol, Thermo Scientific™

Catalog Number p-3523345
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13461-74-0
MFCD00041555
MMZVVJGCZZAWBN-UHFFFAOYSA-N
9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
518771
9-ethynylfluoren-9-ol
C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O
This item is not returnable. View return policy
13461-74-0
MFCD00041555
MMZVVJGCZZAWBN-UHFFFAOYSA-N
9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
518771
9-ethynylfluoren-9-ol
C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O
Melting Point 107.0°C to 110.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C15H10O
Quantity 1 g
Molecular Weight (g/mol) 206.24
Formula Weight 206.24
Percent Purity ≥98.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 9-Ethynyl-9-fluorenol
Show More Show Less
compliance-icons
Product Identifier
  • 9-Ethynyl-9-fluorenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'