missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole-3-boronic acid, 98%, Thermo Scientific™

Catalog Number 15412399
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
669072-93-9
C14H14BNO2
239.08
MFCD09743121
JUBKLYCRFZTXBA-UHFFFAOYSA-N
9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
45382262
CCN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B(O)O
This item is not returnable. View return policy
669072-93-9
C14H14BNO2
239.08
MFCD09743121
JUBKLYCRFZTXBA-UHFFFAOYSA-N
9-ethyl-9h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid, 9-ethyl-9h-carbazol-3-yl boronicacid
45382262
CCN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 23374049
IUPAC Name (9-ethylcarbazol-3-yl)boronic acid
Formula Weight 239.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Ethylcarbazole-3-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'