missing translation for 'onlineSavingsMsg'
Learn More

9-(Chloromethyl)anthracene, 98+%, Thermo Scientific Chemicals

Catalog Number 10499680
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24463-19-2
C15H11Cl
226.7
MFCD00001263
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
32385
9-(chloromethyl)anthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
This item is not returnable. View return policy
24463-19-2
C15H11Cl
226.7
MFCD00001263
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
32385
9-(chloromethyl)anthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 136°C to 141°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 226.7
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 9-(Chloromethyl)anthracene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 9-(Chloromethyl)anthracene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'