missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromofluorene, 98+%, Thermo Scientific Chemicals
21.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
50 g
250 g
Description
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
Specifications
Melting Point | 101°C to 105°C |
Odor | Irritating |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 2047220 |
Solubility Information | Insoluble in water. |
Formula Weight | 245.12 |
Percent Purity | ≥98% |
Chemical Name or Material | 9-Bromofluorene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'