missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromoanthracene, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
9-Bromoanthracene acts as an intermediate in the preparation of 9-substituted derivative of the polycyclic aromatic hydrocarbon (PAH) anthracene.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 97°C to 103°C |
Quantity | 1 g |
Beilstein | 1869747 |
Solubility Information | Insoluble in water. |
Formula Weight | 257.14 |
Percent Purity | 96% |
Chemical Name or Material | 9-Bromoanthracene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'