missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Bromo-10-(2-naphthyl)anthracene, 98%

Brand:  Alfa Aesar™ H64746.MD

Code : NEW

Additional Details : CAS Number : 474688-73-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15422069

  • £10.07 / 250mg


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

9-Bromo-10-(2-naphthyl)anthracene
383.29
10698181
474688-73-8
MFCD09832882

Specifications

9-Bromo-10-(2-naphthyl)anthracene
171° to 173°C
250mg
383.29
10698181
474688-73-8
Solid
98%
MFCD09832882
Videos
SDS