missing translation for 'onlineSavingsMsg'
Learn More

9-Benzhydrylidene-10-anthrone, 95%, Thermo Scientific™

Catalog Number p-63792
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
667-91-4
C27H18O
358.44
MFCD00019172
SPOJSTUMWGMCFP-UHFFFAOYSA-N
69586
10-benzhydrylideneanthracen-9-one
C1=CC=C(C=C1)C(=C2C3=CC=CC=C3C(=O)C4=CC=CC=C42)C5=CC=CC=C5
This item is not returnable. View return policy
667-91-4
C27H18O
358.44
MFCD00019172
SPOJSTUMWGMCFP-UHFFFAOYSA-N
69586
10-benzhydrylideneanthracen-9-one
C1=CC=C(C=C1)C(=C2C3=CC=CC=C3C(=O)C4=CC=CC=C42)C5=CC=CC=C5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 203.0°C to 207.0°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 07, 551
Synonym 9-benzhydrylidene-10-anthrone, 10-diphenylmethylene anthracen-9 10h-one, 10-diphenylmethylidene anthracen-9-one, anthrafuchson, 10-benzhydrylidene-9-anthrone, 10-diphenylmethylene anthracen-9-one, 9-diphenylmethylene-10 9h-anthracenone, 9 10h-anthracenone,10-diphenylmethylene, 10-diphenylmethylene-9 10h-anthracenone #, 9 10h-anthracenone, 10-diphenylmethylene
Formula Weight 358.44
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzhydrylidene-10-anthrone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'