missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-4510260
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
642-31-9
C15H10O
206.24
MFCD00001254
YMNKUHIVVMFOFO-UHFFFAOYSA-N
9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
69504
anthracene-9-carbaldehyde
O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
This item is not returnable. View return policy
642-31-9
C15H10O
206.24
MFCD00001254
YMNKUHIVVMFOFO-UHFFFAOYSA-N
9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
69504
anthracene-9-carbaldehyde
O=CC1=C2C=CC=CC2=CC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103.0°C to 107.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform
Formula Weight 206.24
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 9-Anthracenecarboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'