missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenemethanol, 98%, Thermo Scientific Chemicals

Catalog Number 10502211
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 160.0°C to 164.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 208.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthracenemethanol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'