Alfa Aesar™ 9,9'-Spirobifluorene, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64133.MD

250MG 9,9'-Spirobifluorene, 98% 250mg

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 159-66-0 Weight: 0.00025kg


Quantity 250mg

Product Code. 15461199

Quantity Price
1 £ 22.259999999999998 / 250mg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 9,9'-Spirobifluorene
CAS 159-66-0
Formula Weight 316.4
MDL Number MFCD08704216
Beilstein 1914463

Specification

Formula Weight 316.4
Melting Point 195° to 200°C
Chemical Name or Material 9,9'-Spirobifluorene
Physical Form Solid
Assay 98%
MDL Number MFCD08704216
CAS 159-66-0
Beilstein 1914463
Color White to pale yellow
Quantity 250mg

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.