missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dioctylfluorene-2,7-diboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3759044
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
258865-48-4
C29H44B2O4
478.29
MFCD03701608
HURJMQMZDPOUOU-UHFFFAOYSA-N
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
4283240
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid
CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O
This item is not returnable. View return policy
258865-48-4
C29H44B2O4
478.29
MFCD03701608
HURJMQMZDPOUOU-UHFFFAOYSA-N
9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
4283240
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid
CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 158°C to 162°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 478.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 9, 9-Dioctylfluorene-2, 7-diboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 9,9-Dioctylfluorene-2,7-diboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'