missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester, 97%

Brand:  Alfa Aesar™ H55392.03

Code : NEW

Additional Details : CAS Number : 317802-08-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432086

  • £391.80 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
317802-08-7
558.42
MFCD03701607

Specifications

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
317802-08-7
97%
MFCD03701607
1g
Solid
558.42
Videos
SDS