missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals

Catalog Number p-7037557
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
286438-45-7
C37H56Br2
660.663
MFCD03427215
KFOUJVGPGBSIFB-UHFFFAOYSA-N
2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
5140914
2,7-dibromo-9,9-didodecylfluorene
CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC
This item is not returnable. View return policy
286438-45-7
C37H56Br2
660.663
MFCD03427215
KFOUJVGPGBSIFB-UHFFFAOYSA-N
2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
5140914
2,7-dibromo-9,9-didodecylfluorene
CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC

It is an intermediate for polymeric light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for polymeric light-emitting diodes.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 50°C to 55°C
Boiling Point 180°C (0.15 mmHg)
Flash Point 110°C (230°F)
Quantity 25 g
Formula Weight 660.65
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-dodecyl-2,7-dibromofluorene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'