missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%

Brand:  Alfa Aesar™ H56320.22

Code : NEW

Additional Details : CAS Number : 286438-45-7 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15402788

  • £366.00 / 100g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is an intermediate for polymeric light-emitting diodes.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

286438-45-7
660.663
KFOUJVGPGBSIFB-UHFFFAOYSA-N
5140914
CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC
C37H56Br2
MFCD03427215
2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
2,7-dibromo-9,9-didodecylfluorene

Specifications

9,9-Di-n-dodecyl-2,7-dibromofluorene
180°C (0.15mmHg)
100g
C37H56Br2
2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC
660.663
660.65
110°C (230°F)
50°C to 55°C
286438-45-7
MFCD03427215
KFOUJVGPGBSIFB-UHFFFAOYSA-N
2,7-dibromo-9,9-didodecylfluorene
5140914
98%
Videos
SDS Documents
Certificates