missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 207°C to 212°C |
Density | 1.405 |
Boiling Point | >360°C |
Flash Point | 245°C (473°F) |
Quantity | 5 g |
UN Number | UN3077 |
Beilstein | 608838 |
Merck Index | 14,7213 |
Solubility Information | Insoluble in water. |
Formula Weight | 208.22 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'