missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dibromoanthracene, 96%, Thermo Scientific Chemicals

Catalog Number p-103154
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
523-27-3
C14H8Br2
336.02
MFCD00001244
BRUOAURMAFDGLP-UHFFFAOYSA-N
anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y
68226
9,10-dibromoanthracene
C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br
This item is not returnable. View return policy
523-27-3
C14H8Br2
336.02
MFCD00001244
BRUOAURMAFDGLP-UHFFFAOYSA-N
anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y
68226
9,10-dibromoanthracene
C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 220°C to 225°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05, 665
Merck Index 15, 3023
Formula Weight 336.02
Percent Purity 96%
Physical Form Fluffy Powder
Chemical Name or Material 9, 10-Dibromoanthracene, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 9,10-Dibromoanthracene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'