missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinecarbaldehyde, Thermo Scientific™

Catalog Number p-2703609
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
38707-70-9
C10H7NO
157.172
MFCD00805837
OVZQVGZERAFSPI-UHFFFAOYSA-N
8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
170103
quinoline-8-carbaldehyde
C1=CC2=C(C(=C1)C=O)N=CC=C2
This item is not returnable. View return policy
38707-70-9
C10H7NO
157.172
MFCD00805837
OVZQVGZERAFSPI-UHFFFAOYSA-N
8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
170103
quinoline-8-carbaldehyde
C1=CC2=C(C(=C1)C=O)N=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material 8-Quinolinecarbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'