missing translation for 'onlineSavingsMsg'
Learn More

8-Methylnonanoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-165238
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5963-14-4
C10H19O2
171.26
MFCD00044086
OAOABCKPVCUNKO-UHFFFAOYSA-M
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
111470
8-methylnonanoic acid
CC(C)CCCCCCC([O-])=O
This item is not returnable. View return policy
5963-14-4
C10H19O2
171.26
MFCD00044086
OAOABCKPVCUNKO-UHFFFAOYSA-M
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
111470
8-methylnonanoic acid
CC(C)CCCCCCC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 118.0°C to 120.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CH(CH2)6COOH
Refractive Index 1.4330 to 1.4350
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in thf
Formula Weight 172.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 8-Methylnonanoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Methylnonanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'