missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7036190
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
916162-54-4
C16H20BNO3
285.15
MFCD20527214
UJPLJBDGXPKCDH-UHFFFAOYSA-N
8-methoxyquinoline-5-boronic acid pinacol ester, 8-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 2-8-methoxy-5-quinolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, quinoline, 8-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
71422855
8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CN=C12
This item is not returnable. View return policy
916162-54-4
C16H20BNO3
285.15
MFCD20527214
UJPLJBDGXPKCDH-UHFFFAOYSA-N
8-methoxyquinoline-5-boronic acid pinacol ester, 8-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 2-8-methoxy-5-quinolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, quinoline, 8-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
71422855
8-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CN=C12

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 285.15
Percent Purity 96%
Chemical Name or Material 8-Methoxyquinoline-5-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'