missing translation for 'onlineSavingsMsg'
Learn More

8-M-PDOT, Tocris Bioscience™

Melatonin agonist

Brand:  Tocris (Bio-Techne) 103510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13428213

  • £92.61 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Melatonin receptor agonist, 20-fold selective for the MT2 (vs MT1) subtype. At human recombinant MT1 and MT2 receptors, displays pKi values of 8.23 and 8.95 respectively.
Specifications

Chemical Identifiers

134865-70-6
233.311
8-m-pdot, tocris-1035, n-2r-8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl propanamide
N-[(2R)-8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl]propanamide
C14H19NO2
RVIGBTUDFAGRTQ-LLVKDONJSA-N
6604799
CCC(=O)NC1CCC2=C(C1)C(=CC=C2)OC

Specifications

10mg
134865-70-6
8-m-pdot, tocris-1035, n-2r-8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl propanamide
CCC(=O)NC1CCC2=C(C1)C(=CC=C2)OC
233.311
233.31
8-M-PDOT
C14H19NO2
RVIGBTUDFAGRTQ-LLVKDONJSA-N
N-[(2R)-8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl]propanamide
6604799
>99%
Videos
Documents