missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-147476
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
148-24-3
C9H7NO
145.16
MFCD00006807
MCJGNVYPOGVAJF-UHFFFAOYSA-N
8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
1923
CHEBI:48981
quinolin-8-ol
OC1=C2N=CC=CC2=CC=C1
This item is not returnable. View return policy
148-24-3
C9H7NO
145.16
MFCD00006807
MCJGNVYPOGVAJF-UHFFFAOYSA-N
8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
1923
CHEBI:48981
quinolin-8-ol
OC1=C2N=CC=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 72.0°C to 74.0°C
Color Beige or White to Yellow
Boiling Point 267.0°C (752.0 mmHg)
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, IV, 1135
Merck Index 15, 4881
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,chloroform and,benzene
Formula Weight 145.16
Grade ACS Reagent
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 8-Hydroxyquinoline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'