missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinaldine 98%, ACROS Organics™

Brand:  Acros Organics 121871000

Code : 29

Additional Details : CAS Number : 826-81-3 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10743681

  • £21.80 / 100g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

8-Hydroxyquinaldine
826-81-3
100.0
98%
159.19
2-Methyl-8-quinolinol
98%
97.5
≥97.5%
C10H9NO
MFCD00006765

Specifications

8-Hydroxyquinaldine
68° to 73°C
98%
97.5
Authentic
Crystalline Powder
98%
159.19
21, 106
Beige to brown
100g
826-81-3
100.0
Glass bottle
≥97.5%
C10H9NO
MFCD00006765
2-Methyl-8-quinolinol
Videos
SDS