missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 8-Chlorotheophylline, 99%

Brand:  Alfa Aesar™ A12408.18

Code : NEW

Additional Details : CAS Number : 85-18-7 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11449583

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Applications
8-Chlorotheophylline is a xanthine alkaloid derivative. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in sodium hydroxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

8-Chlorotheophylline
C7H7ClN4O2
MFCD00005581
8-Chloro-1,3-dimethyl-2,6-purinedione
85-18-7
214.61
203068

Specifications

8-Chlorotheophylline
50g
99%
214.61
203068
≈290°C (dec.)
85-18-7
C7H7ClN4O2
MFCD00005581
8-Chloro-1,3-dimethyl-2,6-purinedione
Videos
SDS